siRNA Details
| Gene Symbol: ARL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138450.5 | NP_612459.1 | 2 | 417 | ||
| XM_005266253.3 | XP_005266310.1 | 3 | 326 | |||
| GenBank mRNA | AF441378.1 | - | 2 | 417 | ||
| AK091249.1 | - | 2 | 228 | |||
| BC013150.1 | - | 2 | 196 | 4340470 | ||

ARL11
Genomic Map