siRNA Details
| Gene Symbol: FUS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170634.1 | NP_001164105.1 | 15 | 1739 | ||
| NM_001170937.1 | NP_001164408.1 | 15 | 1730 | |||
| NM_004960.3 | NP_004951.1 | 15 | 1742 | |||
| NR_028388.2 | 14 | 1707 | ||||
| XM_005255233.4 | XP_005255290.1 | 14 | 2370 | |||
| XM_011545781.1 | XP_011544083.1 | 15 | 1708 | |||
| XM_011545782.2 | XP_011544084.1 | 10 | 1098 | |||
| GenBank mRNA | AB208902.1 | - | 13 | 4812 | ||
| AK098774.1 | - | 15 | 1710 | |||
| AK130774.1 | - | 14 | 1679 | |||
| AK290936.1 | - | 15 | 1710 | |||
| AK299129.1 | - | 16 | 1420 | |||
| BC000402.2 | - | 15 | 1705 | 2822692 | ||
| BC002459.1 | - | 15 | 1677 | 3345294 | ||
| BC026062.2 | - | 15 | 1695 | 3937437 | ||
| S62140.1 | - | 15 | 1715 | |||
| X71428.1 | - | 15 | 1706 | |||

FUS
Genomic Map