siRNA Details
| Gene Symbol: ADCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281768.1 | NP_001268697.1 | 9 | 1176 | ||
| NM_021116.2 | NP_066939.1 | 8 | 1490 | |||
| XM_005249584.3 | XP_005249641.1 | 8 | 1488 | |||
| XM_005249585.2 | XP_005249642.1 | 8 | 1488 | |||
| GenBank mRNA | AB209496.1 | - | 8 | 1435 | ||
| AF086306.1 | - | 2 | 92 | |||
| L05500.1 | - | 6 | 634 | |||

ADCY1
Genomic Map