siRNA Details
| Gene Symbol: SLC2A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030777.3 | NP_110404.1 | 3 | 1416 | ||
| XM_011529060.2 | XP_011527362.1 | 3 | 1499 | |||
| XM_011529061.2 | XP_011527363.1 | 3 | 3056 | |||
| XM_011529062.2 | XP_011527364.1 | 3 | 1559 | |||
| XM_011529063.2 | XP_011527365.1 | 3 | 1509 | |||
| XM_011529064.2 | XP_011527366.1 | 3 | 1560 | |||
| XM_011529065.2 | XP_011527367.1 | 3 | 1506 | |||
| XM_017028087.1 | XP_016883576.1 | 3 | 1409 | |||
| GenBank mRNA | AF248053.1 | - | 3 | 1569 | ||
| AF321240.1 | - | 3 | 1371 | |||
| AF479407.1 | - | 2 | 70 | |||
| AK055548.1 | - | 3 | 1416 | |||
| AK290961.1 | - | 3 | 1407 | |||
| AK315269.1 | - | 3 | 1407 | |||
| AL137188.3 | - | 2 | 1327 | |||
| BC101657.1 | - | 3 | 1415 | 8069163 | ||
| BC113423.1 | - | 3 | 1415 | 8322475 | ||

SLC2A10
Genomic Map