siRNA Details
| Gene Symbol: LOXHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145472.2 | NP_001138944.1 | 10 | 1347 | ||
| NM_001308013.1 | NP_001294942.1 | 8 | 1001 | |||
| NM_144612.6 | NP_653213.6 | 28 | 4271 | |||
| XM_006722388.3 | XP_006722451.1 | 9 | 1181 | |||
| XM_006722389.3 | XP_006722452.1 | 10 | 1357 | |||
| XM_006722390.3 | XP_006722453.1 | 10 | 1425 | |||
| XM_006722391.3 | XP_006722454.1 | 9 | 1181 | |||
| XM_011525804.2 | XP_011524106.1 | 16 | 2505 | |||
| XM_011525807.2 | XP_011524109.1 | 7 | 891 | |||
| XM_017025540.1 | XP_016881029.1 | 20 | 3301 | |||
| XM_017025541.1 | XP_016881030.1 | 21 | 3388 | |||
| XM_017025542.1 | XP_016881031.1 | 21 | 3388 | |||
| XM_017025543.1 | XP_016881032.1 | 21 | 3388 | |||
| XM_017025544.1 | XP_016881033.1 | 21 | 3388 | |||
| XM_017025545.1 | XP_016881034.1 | 21 | 3388 | |||
| XM_017025546.1 | XP_016881035.1 | 21 | 3388 | |||
| XM_017025547.1 | XP_016881036.1 | 21 | 3388 | |||
| XM_017025548.1 | XP_016881037.1 | 26 | 3735 | |||
| GenBank mRNA | AK127869.1 | - | 21 | 3584 | ||
| AK302484.1 | - | 9 | 1178 | |||
| AK302848.1 | - | 9 | 1222 | |||
| BX647229.1 | - | 8 | 1001 | |||

LOXHD1
Genomic Map