siRNA Details
| Gene Symbol: NSDHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129765.1 | NP_001123237.1 | 3 | 355 | ||
| NM_015922.2 | NP_057006.1 | 2 | 288 | |||
| XM_011531178.2 | XP_011529480.1 | 4 | 415 | |||
| XM_017029564.1 | XP_016885053.1 | 2 | 220 | |||
| GenBank mRNA | AK026549.1 | - | 2 | 178 | ||
| BC000245.1 | - | 2 | 209 | 3355755 | ||
| BC007816.2 | - | 2 | 259 | 4299819 | ||
| U47105.2 | - | 2 | 288 | |||

NSDHL
Genomic Map