siRNA Details
| Gene Symbol: NDP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000266.3 | NP_000257.1 | 2 | 643 | ||
| GenBank mRNA | AK313409.1 | - | 2 | 422 | ||
| BC029901.1 | - | 1 | 80 | 5179578 | ||
| X65882.1 | - | 2 | 472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDP| Gene Symbol: NDP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000266.3 | NP_000257.1 | 2 | 643 | ||
| GenBank mRNA | AK313409.1 | - | 2 | 422 | ||
| BC029901.1 | - | 1 | 80 | 5179578 | ||
| X65882.1 | - | 2 | 472 | |||