siRNA Details
Gene Symbol: C1orf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276351.1 | NP_001263280.1 | 8 | 1250 | ||
NM_001276352.1 | NP_001263281.1 | 9 | 1276 | |||
NR_075077.1 | 10 | 1419 | ||||
XM_011541465.2 | XP_011539767.1 | 9 | 1368 | |||
XM_011541466.2 | XP_011539768.1 | 9 | 1368 | |||
XM_011541467.1 | XP_011539769.1 | 9 | 1338 | |||
XM_011541469.1 | XP_011539771.1 | 5 | 948 | |||
XM_017001276.1 | XP_016856765.1 | 9 | 1421 | |||
GenBank mRNA | BC047053.1 | - | 5 | 3599 | ||
BC090886.1 | - | 7 | 1222 | 30378691 |
