siRNA Details
| Gene Symbol: AP5Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014855.2 | NP_055670.1 | 17 | 2532 | ||
| XM_017012864.1 | XP_016868353.1 | 16 | 2359 | |||
| GenBank mRNA | AB007875.1 | - | 17 | 2756 | ||
| AK092068.1 | - | 4 | 1995 | |||
| BC008841.2 | - | 8 | 1200 | |||
| BC037399.1 | - | 14 | 2734 | 5172434 | ||

AP5Z1
Genomic Map