siRNA Details
| Gene Symbol: TMEM237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044385.2 | NP_001037850.1 | 12 | 1210 | ||
| NM_152388.3 | NP_689601.2 | 12 | 1375 | |||
| GenBank mRNA | AB053301.1 | - | 12 | 1377 | ||
| AK090601.1 | - | 12 | 1068 | |||
| AK125841.1 | - | 1 | 220 | |||
| AK303954.1 | - | 12 | 1197 | |||
| AK304395.1 | - | 10 | 899 | |||
| BC013730.1 | - | 4 | 293 | |||
| BC029611.1 | - | 12 | 1123 | 5304046 | ||
| BX538000.1 | - | 4 | 270 | |||

TMEM237
Genomic Map