siRNA Details
| Gene Symbol: TMEM237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044385.2 | NP_001037850.1 | 9 | 934 | ||
| NM_152388.3 | NP_689601.2 | 9 | 1099 | |||
| GenBank mRNA | AB053301.1 | - | 9 | 1101 | ||
| AF212043.1 | - | 4 | 503 | |||
| AK090601.1 | - | 9 | 792 | |||
| AK303954.1 | - | 9 | 921 | |||
| AK304395.1 | - | 7 | 623 | |||
| BC013730.1 | - | 1 | 17 | |||
| BC029611.1 | - | 9 | 847 | 5304046 | ||

TMEM237
Genomic Map