siRNA Details
| Gene Symbol: SPAST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014946.3 | NP_055761.2 | 2 | 673 | ||
| NM_199436.1 | NP_955468.1 | 2 | 673 | |||
| XM_005264516.4 | XP_005264573.1 | 2 | 449 | |||
| XM_011533067.2 | XP_011531369.1 | 2 | 733 | |||
| XM_017004777.1 | XP_016860266.1 | 2 | 730 | |||
| XM_017004778.1 | XP_016860267.1 | 2 | 733 | |||
| GenBank mRNA | AB029006.1 | - | 2 | 673 | ||
| AJ246001.1 | - | 2 | 577 | |||
| BC150260.1 | - | 2 | 673 | 9007202 | ||

SPAST
Genomic Map