siRNA Details
| Gene Symbol: AMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030943.3 | NP_112205.2 | 2 | 127 | ||
| XM_011537202.2 | XP_011535504.1 | 2 | 372 | |||
| XM_011537203.2 | XP_011535505.1 | 2 | 628 | |||
| GenBank mRNA | AF328788.1 | - | 2 | 116 | ||
| AK055802.1 | - | 1 | 840 | |||
| AY358468.1 | - | 2 | 114 | |||
| BC029948.1 | - | 2 | 127 | |||

AMN
Genomic Map