siRNA Details
| Gene Symbol: SLC29A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174098.1 | NP_001167569.1 | 6 | 1683 | ||
| NM_018344.5 | NP_060814.4 | 6 | 1677 | |||
| NR_033413.1 | 5 | 1594 | ||||
| NR_033414.1 | 4 | 1367 | ||||
| XM_006717910.3 | XP_006717973.1 | 6 | 2069 | |||
| XM_017016377.1 | XP_016871866.1 | 5 | 1891 | |||
| XM_017016378.1 | XP_016871867.1 | 4 | 1667 | |||
| GenBank mRNA | AK002022.1 | - | 5 | 1428 | ||
| AK304503.1 | - | 5 | 1565 | |||
| AX775915.1 | - | 6 | 1677 | |||
| AX775917.1 | - | 6 | 1677 | |||
| AX775919.1 | - | 6 | 1677 | |||
| AX775921.1 | - | 6 | 1677 | |||
| AY358686.1 | - | 6 | 1655 | |||
| BC000223.1 | - | 3 | 1052 | |||
| BC041575.1 | - | 5 | 1565 | |||

SLC29A3
Genomic Map