siRNA Details
| Gene Symbol: FGF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291812.1 | NP_001278741.1 | 3 | 582 | ||
| NM_004464.3 | NP_004455.2 | 3 | 870 | |||
| NM_033143.2 | NP_149134.1 | 2 | 766 | |||
| GenBank mRNA | AB016517.1 | - | 2 | 529 | ||
| AF171928.1 | - | 2 | 679 | |||
| AF535149.1 | - | 3 | 795 | |||
| AK090557.1 | - | 3 | 582 | |||
| AK291962.1 | - | 3 | 795 | |||
| BC074858.2 | - | 3 | 682 | 30915226 | ||
| BC074859.2 | - | 3 | 682 | 30915463 | ||
| BC131502.1 | - | 3 | 771 | 40082281 | ||
| M37825.1 | - | 3 | 772 | |||

FGF5
Genomic Map