siRNA Details
Gene Symbol: EFNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004429.4 | NP_004420.1 | 5 | 1420 | ||
GenBank mRNA | BC016649.1 | - | 5 | 1420 | 3867060 | |
BC052979.1 | - | 5 | 1439 | 6164440 | ||
L37361.1 | - | 5 | 1166 | |||
U09303.1 | - | 5 | 1341 | |||
U09304.1 | - | 5 | 947 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
