siRNA Details
| Gene Symbol: MESP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039958.1 | NP_001035047.1 | 2 | 1303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MESP2| Gene Symbol: MESP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039958.1 | NP_001035047.1 | 2 | 1303 | ||