siRNA Details
| Gene Symbol: RBM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204466.1 | NP_001191395.1 | 20 | 2577 | ||
| NM_001204467.1 | NP_001191396.1 | 21 | 2805 | |||
| NM_001204468.1 | NP_001191397.1 | 21 | 2839 | |||
| NM_005676.4 | NP_005667.2 | 21 | 2808 | |||
| NM_152856.2 | NP_690595.1 | 20 | 2574 | |||
| XM_005272677.4 | XP_005272734.1 | 21 | 2858 | |||
| XM_005272678.4 | XP_005272735.1 | 20 | 2392 | |||
| XM_005272679.4 | XP_005272736.1 | 20 | 2389 | |||
| XM_006724563.1 | XP_006724626.1 | 17 | 2025 | |||
| XM_011543989.2 | XP_011542291.1 | 13 | 1749 | |||
| XM_017029884.1 | XP_016885373.1 | 20 | 2728 | |||
| XM_017029885.1 | XP_016885374.1 | 20 | 2500 | |||
| GenBank mRNA | AK024839.1 | - | 4 | 416 | ||
| AK025854.1 | - | 9 | 1181 | |||
| AK098115.1 | - | 2143 | ||||
| AK225175.1 | - | 12 | 1480 | |||
| AK292758.1 | - | 20 | 2528 | |||
| AL137421.1 | - | 22 | 2697 | |||
| BC003089.1 | - | 20 | 2513 | 3536058 | ||
| BC004181.2 | - | 21 | 2761 | 2961094 | ||
| BC008733.2 | - | 21 | 2793 | 3163064 | ||
| BC024153.2 | - | 20 | 2423 | 3845258 | ||
| BX537969.1 | - | 21 | 2790 | |||
| D50912.1 | - | 21 | 2656 | |||
| KU178411.1 | - | 19 | 2181 | |||
| KU178412.1 | - | 20 | 2415 | |||
| U35373.1 | - | 7 | 792 | |||

RBM10
Genomic Map