siRNA Details
| Gene Symbol: SGCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000337.5 | NP_000328.2 | 6 | 919 | ||
| NM_001128209.1 | NP_001121681.1 | 5 | 873 | |||
| NM_172244.2 | NP_758447.1 | 6 | 919 | |||
| XM_005265966.4 | XP_005266023.1 | 6 | 747 | |||
| XM_005265967.2 | XP_005266024.1 | 6 | 511 | |||
| XM_011534621.2 | XP_011532923.1 | 5 | 576 | |||
| XM_017009723.1 | XP_016865212.1 | 7 | 685 | |||
| XM_017009724.1 | XP_016865213.1 | 7 | 962 | |||
| GenBank mRNA | AK292794.1 | - | 6 | 511 | ||
| BC020740.1 | - | 6 | 512 | 4702959 | ||
| BX537948.1 | - | 5 | 884 | |||
| DA561069.1 | - | 5 | 466 | |||
| U58331.1 | - | 5 | 438 | |||
| X95191.1 | - | 4 | 400 | |||

SGCD
Genomic Map