siRNA Details
| Gene Symbol: RSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286506.1 | NP_001273435.1 | 6 | 673 | ||
| NM_080860.3 | NP_543136.1 | 7 | 787 | |||
| XM_005261208.2 | XP_005261265.1 | 5 | 427 | |||
| XM_011529786.1 | XP_011528088.1 | 6 | 607 | |||
| GenBank mRNA | AB006536.1 | - | 7 | 787 | ||
| AB041016.1 | - | 7 | 724 | |||
| AK057315.1 | - | 6 | 2238 | |||
| AK314746.1 | - | 7 | 624 | |||
| BC101519.1 | - | 7 | 666 | 8069025 | ||
| BC113367.1 | - | 7 | 666 | 8322419 | ||
| BG822861.1 | - | 507 | ||||

RSPH1
Genomic Map