siRNA Details
| Gene Symbol: NBPF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037675.3 | NP_001032764.2 | 13 | 1773 | ||
| NM_001277444.1 | NP_001264373.1 | 13 | 1773 | |||
| XM_006711317.2 | XP_006711380.1 | 14 | 2001 | |||
| GenBank mRNA | AF419618.1 | - | 6 | 721 | ||
| AK095030.1 | - | 1 | 1006 | |||
| AK294414.1 | - | 7 | 931 | |||
| AY894574.1 | - | 13 | 1775 | |||
| BC071995.1 | - | 8 | 957 | |||
| BC094841.1 | - | 8 | 957 | |||

NBPF9
Genomic Map