siRNA Details
| Gene Symbol: CHMP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244644.1 | NP_001231573.1 | 2 | 478 | ||
| NM_014043.3 | NP_054762.2 | 3 | 570 | |||
| XM_011533576.2 | XP_011531878.1 | 3 | 401 | |||
| GenBank mRNA | BI821848.1 | - | 3 | 547 | ||
| DA426159.1 | - | 3 | 541 | |||

CHMP2B
Genomic Map