siRNA Details
| Gene Symbol: C20orf202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009612.2 | NP_001009612.1 | 2 | 1003 | ||
| GenBank mRNA | AK092524.1 | - | 2 | 1003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf202| Gene Symbol: C20orf202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009612.2 | NP_001009612.1 | 2 | 1003 | ||
| GenBank mRNA | AK092524.1 | - | 2 | 1003 | ||