siRNA Details
| Gene Symbol: C20orf202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009612.2 | NP_001009612.1 | 1 | 112 | ||
| GenBank mRNA | AK092524.1 | - | 1 | 112 | ||
| BC127730.1 | - | 1 | 42 | |||
| BC127731.1 | - | 1 | 42 | |||
| BC141955.1 | - | 1 | 42 | |||
| BC141956.1 | - | 1 | 42 | |||
| BC150532.1 | - | 1 | 42 | |||
| CR746175.1 | - | 1 | 54 | |||

C20orf202
Genomic Map