siRNA Details
| Gene Symbol: ERLIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007175.6 | NP_009106.1 | 7 | 574 | ||
| XM_005273392.2 | XP_005273449.1 | 7 | 734 | |||
| XM_006716280.2 | XP_006716343.1 | 6 | 479 | |||
| GenBank mRNA | AK291394.1 | - | 7 | 527 | ||
| AL442077.1 | - | 561 | ||||
| AY358108.1 | - | 7 | 518 | |||
| BC005950.1 | - | 7 | 497 | |||
| BP353279.1 | - | 7 | 574 | |||

ERLIN2
Genomic Map