siRNA Details
| Gene Symbol: HPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171993.1 | NP_001165464.1 | 11 | 1058 | ||
| NM_002150.2 | NP_002141.1 | 9 | 582 | |||
| GenBank mRNA | AK057510.1 | - | 11 | 1058 | ||
| AK290826.1 | - | 9 | 578 | |||
| BC024287.1 | - | 9 | 564 | 5087393 | ||
| X72389.1 | - | 9 | 567 | |||

HPD
Genomic Map