siRNA Details
| Gene Symbol: HPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171993.1 | NP_001165464.1 | 15, 16 | 1586 | ||
| NM_002150.2 | NP_002141.1 | 13, 14 | 1110 | |||
| GenBank mRNA | AK057510.1 | - | 15, 16 | 1586 | ||
| AK290826.1 | - | 13, 14 | 1106 | |||
| BC024287.1 | - | 13, 14 | 1092 | 5087393 | ||
| X72389.1 | - | 13, 14 | 1095 | |||

HPD
Genomic Map