siRNA Details
| Gene Symbol: RBM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134363.2 | NP_001127835.2 | 4 | 1416 | ||
| XM_011539697.2 | XP_011537999.1 | 4 | 1254 | |||
| XM_017016103.1 | XP_016871592.1 | 4 | 2674 | |||
| XM_017016104.1 | XP_016871593.1 | 4 | 4890 | |||
| GenBank mRNA | EU822950.1 | - | 4 | 1416 | ||

RBM20
Genomic Map