siRNA Details
| Gene Symbol: PDSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020381.3 | NP_065114.3 | 6 | 1219 | ||
| XM_011535956.2 | XP_011534258.1 | 6 | 1317 | |||
| XM_011535958.2 | XP_011534260.1 | 5 | 1183 | |||
| XM_011535960.2 | XP_011534262.1 | 6 | 946 | |||
| XM_011535962.2 | XP_011534264.1 | 6 | 861 | |||
| XM_017011082.1 | XP_016866571.1 | 6 | 1239 | |||
| GenBank mRNA | AB210839.1 | - | 6 | 929 | ||
| AF254956.1 | - | 6 | 1190 | |||
| AK225629.1 | - | 6 | 1200 | |||
| AK301480.1 | - | 6 | 822 | |||
| AK315804.1 | - | 6 | 1200 | |||
| AL832290.1 | - | 1 | 24 | |||
| BC039906.1 | - | 6 | 1205 | 6066481 | ||

PDSS2
Genomic Map