siRNA Details
| Gene Symbol: WDR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006657.1 | NP_001006658.1 | 5 | 443 | ||
| NM_020779.3 | NP_065830.2 | 5 | 443 | |||
| GenBank mRNA | AB037757.2 | - | 5 | 451 | ||
| AK122917.1 | - | 5 | 368 | |||
| AK315094.1 | - | 5 | 368 | |||
| BC036659.2 | - | 5 | 442 | 5266940 | ||

WDR35
Genomic Map