siRNA Details
| Gene Symbol: WDR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006657.1 | NP_001006658.1 | 19 | 2096 | ||
| NM_020779.3 | NP_065830.2 | 18 | 2063 | |||
| XM_011533007.2 | XP_011531309.1 | 8 | 788 | |||
| GenBank mRNA | AB037757.2 | - | 19 | 2104 | ||
| AK091205.1 | - | 12 | 1431 | |||
| AK122917.1 | - | 18 | 1988 | |||
| AK315094.1 | - | 18 | 1988 | |||
| AL832903.1 | - | 7 | 593 | |||
| BC036659.2 | - | 18 | 2062 | 5266940 | ||

WDR35
Genomic Map