siRNA Details
| Gene Symbol: LINC01376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135287.1 | 2 | 474 | |||
| GenBank mRNA | AK123475.1 | - | 2 | 476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01376| Gene Symbol: LINC01376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135287.1 | 2 | 474 | |||
| GenBank mRNA | AK123475.1 | - | 2 | 476 | ||