siRNA Details
| Gene Symbol: C2orf91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242815.1 | NP_001229744.1 | 2 | 376 | ||
| XM_017004097.1 | XP_016859586.1 | 2 | 6859 | |||
| GenBank mRNA | AK124893.1 | - | 2 | 376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf91| Gene Symbol: C2orf91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242815.1 | NP_001229744.1 | 2 | 376 | ||
| XM_017004097.1 | XP_016859586.1 | 2 | 6859 | |||
| GenBank mRNA | AK124893.1 | - | 2 | 376 | ||