siRNA Details
| Gene Symbol: FTCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006723961.3 | XP_006724024.2 | 11 | 1708 | ||
| XM_006723962.3 | XP_006724025.2 | 11 | 2316 | |||
| XM_011529434.2 | XP_011527736.1 | 11 | 2331 | |||
| XM_011529436.2 | XP_011527738.1 | 11 | 2316 | |||
| XM_011529437.2 | XP_011527739.1 | 11 | 1710 | |||
| XM_011529439.2 | XP_011527741.1 | 9 | 1120 | |||
| GenBank mRNA | AL109817.1 | - | 1 | 324 | ||

FTCD
Genomic Map