siRNA Details
| Gene Symbol: ARX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139058.2 | NP_620689.1 | 1 | 261 | ||
| GenBank mRNA | AY038071.1 | - | 1 | 50 | ||
| BQ100952.1 | - | 1 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARX| Gene Symbol: ARX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139058.2 | NP_620689.1 | 1 | 261 | ||
| GenBank mRNA | AY038071.1 | - | 1 | 50 | ||
| BQ100952.1 | - | 1 | 261 | |||