siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000065.3 | NP_000056.2 | 13 | 2114 | ||
| NM_001115131.2 | NP_001108603.2 | 13 | 2141 | |||
| XM_005248357.2 | XP_005248414.1 | 13 | 2189 | |||
| XM_006714496.3 | XP_006714559.1 | 14 | 2246 | |||
| XM_011514114.2 | XP_011512416.1 | 14 | 2246 | |||
| XM_011514115.2 | XP_011512417.1 | 14 | 2167 | |||
| XM_011514116.2 | XP_011512418.1 | 15 | 2283 | |||
| XM_011514117.2 | XP_011512419.1 | 15 | 2325 | |||
| XM_011514118.2 | XP_011512420.1 | 14 | 2063 | |||
| XM_011514119.2 | XP_011512421.1 | 13 | 2189 | |||
| XM_011514121.2 | XP_011512423.1 | 9 | 1332 | |||
| XM_017009818.1 | XP_016865307.1 | 15 | 2306 | |||
| XM_017009819.1 | XP_016865308.1 | 10 | 1509 | |||

C6
Genomic Map