siRNA Details
| Gene Symbol: TMEM240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114748.1 | NP_001108220.1 | 2 | 110 | ||
| GenBank mRNA | BC147018.1 | - | 3 | 139 | 9056946 | |
| BC147020.1 | - | 3 | 139 | 9056948 | ||
| BU929336.1 | - | 2 | 91 | |||

TMEM240
Genomic Map