siRNA Details
Gene Symbol: DLX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005221.5 | NP_005212.1 | 3 | 778 | ||
XM_005250185.3 | XP_005250242.1 | 3 | 1889 | |||
XM_017011803.1 | XP_016867292.1 | 3 | 279 | |||
GenBank mRNA | AK023493.1 | - | 3 | 778 | ||
BC006226.2 | - | 3 | 764 | 3941691 | ||
BT006903.1 | - | 3 | 570 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AF033011, AK134920, AF022077, U67840, AF022075, and others.
Panther Classification:
Gene Ontology Categories:
