siRNA Details
| Gene Symbol: TBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005996.3 | NP_005987.3 | 2 | 1375 | ||
| NM_016569.3 | NP_057653.3 | 2 | 1375 | |||
| GenBank mRNA | AF002228.1 | - | 2 | 411 | ||
| AF170708.2 | - | 2 | 671 | |||
| AF216750.1 | - | 2 | 527 | |||
| AK054604.1 | - | 2 | 1378 | |||
| AK304639.1 | - | 2 | 645 | |||
| BC025258.1 | - | 2 | 801 | 5458526 | ||

TBX3
Genomic Map