siRNA Details
| Gene Symbol: PDE6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000283.3 | NP_000274.2 | 17 | 2087 | ||
| NM_001145291.1 | NP_001138763.1 | 17 | 2087 | |||
| NM_001145292.1 | NP_001138764.1 | 15 | 1466 | |||
| XM_011513473.2 | XP_011511775.1 | 18 | 3354 | |||
| XM_011513474.2 | XP_011511776.1 | 18 | 3354 | |||
| XM_011513475.2 | XP_011511777.1 | 20 | 3457 | |||
| XM_011513476.2 | XP_011511778.1 | 18 | 3354 | |||
| XM_011513477.2 | XP_011511779.1 | 13 | 2206 | |||
| XM_011513478.2 | XP_011511780.1 | 14 | 2273 | |||
| XM_017008283.1 | XP_016863772.1 | 15 | 1493 | |||
| XM_017008284.1 | XP_016863773.1 | 15 | 1448 | |||
| XM_017008285.1 | XP_016863774.1 | 15 | 1619 | |||
| XM_017008286.1 | XP_016863775.1 | 15 | 1448 | |||
| XM_017008287.1 | XP_016863776.1 | 14 | 1549 | |||
| XM_017008288.1 | XP_016863777.1 | 12 | 2158 | |||
| XM_017008289.1 | XP_016863778.1 | 13 | 1346 | |||
| GenBank mRNA | AK295285.1 | - | 15 | 1463 | ||
| AK316054.1 | - | 15 | 1423 | |||
| BC000249.1 | - | 17 | 2077 | 3356353 | ||
| BT009794.1 | - | 17 | 2034 | |||
| S41458.1 | - | 17 | 2055 | |||
| X66142.1 | - | 17 | 2039 | |||

PDE6B
Genomic Map