siRNA Details
| Gene Symbol: NUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138459.3 | NP_612468.1 | 2 | 707 | ||
| GenBank mRNA | AK125119.1 | - | 2 | 669 | ||
| AK126335.1 | - | 3 | 421 | |||
| AK130093.1 | - | 1 | 24 | |||
| BC013026.2 | - | 2 | 654 | 3531630 | ||
| BC063794.1 | - | 2 | 661 | 6500832 | ||
| BC066910.1 | - | 2 | 701 | 4830860 | ||
| BC110325.1 | - | 2 | 663 | 5553827 | ||
| BC150654.1 | - | 2 | 674 | 9057025 | ||
| BC150655.1 | - | 2 | 674 | 9057026 | ||

NUS1
Genomic Map