siRNA Details
| Gene Symbol: UGT2B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001074.2 | NP_001065.2 | 1 | 183 | ||
| XM_011532229.2 | XP_011530531.1 | 1 | 184 | |||
| XM_011532230.1 | XP_011530532.1 | 1 | 184 | |||
| GenBank mRNA | AK223142.1 | - | 1 | 157 | ||
| AK313190.1 | - | 1 | 184 | |||
| BC030974.1 | - | 1 | 183 | 4593426 | ||
| J05428.1 | - | 1 | 151 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BP277631.1 | - | 1 | 174 | Chr. 4 - 69096494 - 69097045 [+] on Build GRCh38 | |

UGT2B7
Genomic Map