siRNA Details
| Gene Symbol: DGUOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318860.1 | NP_001305789.1 | 2, 3 | 424 | ||
| NM_001318861.1 | NP_001305790.1 | 3, 4 | 510 | |||
| NM_001318862.1 | NP_001305791.1 | 3, 4 | 492 | |||
| NM_001318863.1 | NP_001305792.1 | 2, 3 | 406 | |||
| NM_080916.2 | NP_550438.1 | 3, 4 | 537 | |||
| NR_134894.1 | 2, 3 | 424 | ||||
| XM_011532647.2 | XP_011530949.1 | 3, 4 | 515 | |||
| GenBank mRNA | BC015757.1 | - | 3, 4 | 476 | 4853652 | |
| BF975904.1 | - | 2, 3 | 351 | |||
| BG470729.1 | - | 2, 3 | 340 | |||
| BG575800.1 | - | 3, 4 | 415 | |||
| BG708680.1 | - | 2, 3 | 413 | |||
| BG715600.1 | - | 2, 3 | 365 | |||
| CN311954.1 | - | 3, 4 | 449 | |||
| KU178023.1 | - | 3, 4 | 452 | |||
| KU178024.1 | - | 2, 3 | 339 | |||
| U41668.1 | - | 3, 4 | 486 | |||

DGUOK
Genomic Map