siRNA Details
| Gene Symbol: CSTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005213.3 | NP_005204.1 | 2 | 230 | ||
| GenBank mRNA | AK291308.1 | - | 2 | 146 | ||
| AV716538.1 | - | 2 | 244 | |||
| AV717631.1 | - | 2 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSTA| Gene Symbol: CSTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005213.3 | NP_005204.1 | 2 | 230 | ||
| GenBank mRNA | AK291308.1 | - | 2 | 146 | ||
| AV716538.1 | - | 2 | 244 | |||
| AV717631.1 | - | 2 | 140 | |||