siRNA Details
| Gene Symbol: DLL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019074.3 | NP_061947.1 | 9 | 1831 | ||
| GenBank mRNA | AB036931.1 | - | 9 | 1771 | ||
| AB043894.1 | - | 9 | 1495 | |||
| AF253468.1 | - | 9 | 1495 | |||
| AF279305.1 | - | 8 | 1401 | |||
| AK313831.1 | - | 9 | 1826 | |||
| AY358894.1 | - | 9 | 1530 | |||
| BC106950.2 | - | 9 | 1631 | 40034887 | ||

DLL4
Genomic Map