siRNA Details
| Gene Symbol: FSIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173651.3 | NP_775922.3 | 17 | 16799 | ||
| GenBank mRNA | AK126051.1 | - | 1 | 305 | ||
| BX647691.1 | - | 1 | 24 | |||
| BX648733.1 | - | 1 | 1072 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FSIP2| Gene Symbol: FSIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173651.3 | NP_775922.3 | 17 | 16799 | ||
| GenBank mRNA | AK126051.1 | - | 1 | 305 | ||
| BX647691.1 | - | 1 | 24 | |||
| BX648733.1 | - | 1 | 1072 | |||