siRNA Details
| Gene Symbol: PEX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004813.2 | NP_004804.1 | 9 | 1093 | ||
| NM_057174.2 | NP_476515.1 | 9 | 1093 | |||
| XM_011520474.1 | XP_011518776.1 | 9 | 832 | |||
| GenBank mRNA | AB016531.1 | - | 9 | 818 | ||
| AF118240.1 | - | 9 | 1025 | |||
| BC000467.2 | - | 9 | 799 | 2822827 | ||
| BC004356.1 | - | 9 | 791 | 2822311 | ||

PEX16
Genomic Map