siRNA Details
| Gene Symbol: PEX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004813.2 | NP_004804.1 | 11 | 1591 | ||
| NM_057174.2 | NP_476515.1 | 11 | 1456 | |||
| GenBank mRNA | BC000467.2 | - | 11 | 1162 | 2822827 | |
| BM746616.1 | - | 3 | 411 | |||

PEX16
Genomic Map