siRNA Details
| Gene Symbol: RAB23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278666.1 | NP_001265595.1 | 7 | 3302 | ||
| NM_001278667.1 | NP_001265596.1 | 7 | 3474 | |||
| NM_001278668.1 | NP_001265597.1 | 7 | 3301 | |||
| NM_016277.4 | NP_057361.3 | 7 | 3771 | |||
| NM_183227.2 | NP_899050.1 | 7 | 3325 | |||
| NR_103822.1 | 4 | 2976 | ||||

RAB23
Genomic Map