siRNA Details
| Gene Symbol: LOC401068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028186.1 | - | 1 | 1007 | ||
| Gene Symbol: LOC105377135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534353.2 | XP_011532655.1 | 3 | 1877 | ||

LOC401068
Genomic Map